missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Aminobutyric acid, 99%

Catalog No. AAL0603514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0603514 25 g
AAL0603522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0603514 Supplier Thermo Scientific Chemicals Supplier No. L0603514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2835-81-6 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2835-81-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008093
InChI Key QWCKQJZIFLGMSD-UHFFFAOYSA-N
Synonym dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
PubChem CID 6657
ChEBI CHEBI:35621
IUPAC Name 2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Melting Point ∽291°C (decomposition)
Quantity 25 g
Beilstein 635889
Merck Index 14,428
Formula Weight 103.12
Percent Purity 99%
Chemical Name or Material DL-2-Aminobutyric acid
EINECSNumber 220-616-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only