missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Aminoadipic acid, 98%

CAS: 542-32-5 | C6H11NO4 | 161.16 g/mol

$172.84 - $487.22

Chemical Identifiers

CAS 542-32-5
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00063119,MFCD00063118
InChI Key OYIFNHCXNCRBQI-UHFFFAOYNA-N
Synonym dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
PubChem CID 469
ChEBI CHEBI:37024
IUPAC Name 2-aminohexanedioic acid
SMILES NC(CCCC(O)=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1271703
View Documents
Thermo Scientific Chemicals
L1271703
1 g
Each for $172.84
Only null left
 
AAL1271706
View Documents
Thermo Scientific Chemicals
L1271706
5 g
Each for $487.22
Only null left
 
Description

Description

DL-2-Aminoadipic acid is a labeled amino acid which is used in chemical research and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-2-Aminoadipic acid is a labeled amino acid which is used in chemical research and as pharmaceutical intermediate.

Solubility
Soluble in water (2.2 mg/ml at 20°C), 1 M HCl (50 mg/ml), ethanol (slightly), ether (slightly), and 80% formic acid (25 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Keep in a dry place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

542-32-5
161.16
OYIFNHCXNCRBQI-UHFFFAOYNA-N
469
2-aminohexanedioic acid
C6H11NO4
MFCD00063119,MFCD00063118
dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
CHEBI:37024
NC(CCCC(O)=O)C(O)=O

Specifications

542-32-5
C6H11NO4
1 g
14,416
Soluble in water (2.2mg/ml at 20°C),1 M HCl (50mg/ml),ethanol (slightly),ether (slightly),and 80% formic acid (25mg/ml).
NC(CCCC(O)=O)C(O)=O
161.16
CHEBI:37024
98%
196°C to 198°C
MFCD00063119,MFCD00063118
1773077
dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
OYIFNHCXNCRBQI-UHFFFAOYNA-N
2-aminohexanedioic acid
469
161.16
DL-2-Aminoadipic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-809-2

RTECSNumber : MO1852000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only