missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-1-propanol 98.0+%, TCI America™

Catalog No. p-7117143
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09425G 5 g
A094225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09425G Supplier TCI America Supplier No. A09425G
Only null left

Chemical Identifiers

CAS 6168-72-5
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008085
InChI Key BKMMTJMQCTUHRP-UHFFFAOYNA-N
Synonym dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine
PubChem CID 5126
IUPAC Name 2-aminopropan-1-ol
SMILES CC(N)CO

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
UN Number 2735
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material DL-2-Amino-1-propanol
EINECSNumber (2)-0323&(9)-0122
TSCA Yes