missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Amino-1-butanol 98.0+%, TCI America™
$41.02 - $212.51
Chemical Identifiers
| CAS | 96-20-8 |
|---|---|
| Molecular Formula | C4H11NO |
| Molecular Weight (g/mol) | 89.138 |
| MDL Number | MFCD00008095 |
| InChI Key | JCBPETKZIGVZRE-UHFFFAOYSA-N |
| Synonym | 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol |
| PubChem CID | 22129 |
| IUPAC Name | 2-aminobutan-1-ol |
| SMILES | CCC(CO)N |
Chemical Identifiers
| 96-20-8 | |
| 89.138 | |
| JCBPETKZIGVZRE-UHFFFAOYSA-N | |
| 22129 | |
| CCC(CO)N |
| C4H11NO | |
| MFCD00008095 | |
| 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol | |
| 2-aminobutan-1-ol |
Specifications
| 96-20-8 | |
| Yellow | |
| C4H11NO | |
| 25 g | |
| 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol | |
| CCC(CO)N | |
| 89.138 | |
| 89.14 | |
| Liquid |
| 2°C | |
| 180°C | |
| MFCD00008095 | |
| 3267 | |
| JCBPETKZIGVZRE-UHFFFAOYSA-N | |
| 2-aminobutan-1-ol | |
| 22129 | |
| ≥98.0% (GC,T) | |
| DL-2-Amino-1-butanol |
Safety and Handling
EINECSNumber : (2)-0316
RTECSNumber : EK9625000
TSCA : Yes