missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Amino-1-butanol 98.0+%, TCI America™

Catalog No. A0684500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0684500G 500 g
A068425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0684500G Supplier TCI America Supplier No. A0684500G

Chemical Identifiers

CAS 96-20-8
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008095
InChI Key JCBPETKZIGVZRE-UHFFFAOYSA-N
Synonym 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol
PubChem CID 22129
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

Melting Point 2°C
Color Yellow
Boiling Point 180°C
Quantity 500 g
UN Number 3267
Formula Weight 89.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material DL-2-Amino-1-butanol
EINECSNumber (2)-0316
RTECSNumber EK9625000
TSCA Yes