missing translation for 'onlineSavingsMsg'
Learn More

DL-2,4-Diaminobutyric Acid Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7126627
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11751G 1 g
D11755G 5 g
D117525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D11751G Supplier TCI America Supplier No. D11751G
Only null left

Chemical Identifiers

CAS 65427-54-5
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00012910
InChI Key CKAAWCHIBBNLOJ-UHFFFAOYSA-N
Synonym dl-2,4-diaminobutyric acid dihydrochloride, 2,4-diaminobutanoic acid dihydrochloride, h-dl-dab.2hcl, l-2,4-diaminobutyric acid dihydrochloride, dl-2,4-diaminobutyricaciddihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, l-2,4-diaminobutyric aicd dihydrochloride, 2,4-bis azanyl butanoic acid dihydrochloride, c4h10n2o2.2hcl, h-dl-dab-oh.2hcl
PubChem CID 2724329
IUPAC Name 2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl

Specifications

Melting Point 203°C
Color White
Quantity 1 g
Formula Weight 191.05
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2,4-Diaminobutyric Acid Dihydrochloride