missing translation for 'onlineSavingsMsg'
Learn More

DL-2,4-Diaminobutyric acid dihydrochloride, 99%

Catalog No. AAA1540803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1540803 1 g
AAA1540806 5 g
AAA1540814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1540803 Supplier Thermo Scientific Chemicals Supplier No. A1540803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 65427-54-5 | C4H12Cl2N2O2 | 191.052 g/mol

2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activityagainst mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Diaminobutyric Acid is also found to inhibit GABA transaminase, thus producing an elevation of GABA level, and to have an antitumor activity against mouse fibrosarcoma cells. It is also used as an internal standard for amino acid analysis.

Solubility
Soluble in water 50 mg/mL.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 65427-54-5
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00012910
InChI Key CKAAWCHIBBNLOJ-UHFFFAOYSA-N
Synonym dl-2,4-diaminobutyric acid dihydrochloride, 2,4-diaminobutanoic acid dihydrochloride, h-dl-dab.2hcl, l-2,4-diaminobutyric acid dihydrochloride, dl-2,4-diaminobutyricaciddihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, l-2,4-diaminobutyric aicd dihydrochloride, 2,4-bis azanyl butanoic acid dihydrochloride, c4h10n2o2.2hcl, h-dl-dab-oh.2hcl
PubChem CID 2724329
IUPAC Name 2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl

Specifications

Melting Point ∼198°C (decomposition)
Quantity 1 g
Beilstein 4708499
Sensitivity Hygroscopic
Solubility Information Soluble in water 50mg/mL.
Formula Weight 191.06
Percent Purity 99%
Chemical Name or Material DL-2,4-Diaminobutyric acid dihydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 265-774-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only