missing translation for 'onlineSavingsMsg'
Learn More

DL-2,3-Diaminopropionic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7126213
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
D04381G 1 g
D04385G 5 g
D043825G 25 g
3 options

Catalog No. D04381G

Supplier: TCI America D04381G

Only null left

Chemical Identifiers

CAS 54897-59-5
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00012884
InChI Key SKWCZPYWFRTSDD-UHFFFAOYSA-N
Synonym 2,3-diaminopropanoic acid hydrochloride, dl-2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine hydrochloride, dl-2,3-diaminopropionic acid monohydrochloride, 2,3-diaminopropionic acid hydrochloride, +--2,3-diaminopropionic acid hydrochloride, 3-amino-dl-alanine monohydrochloride, 2,3-diaminopropionic acid monohydrochloride, dl-2,3-diaminopropanoic acid hydrochloride, diaminopropionic acid
PubChem CID 108638
IUPAC Name 2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 237°C
Color White-Yellow
Quantity 1 g
Formula Weight 140.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material DL-2,3-Diaminopropionic Acid Hydrochloride