missing translation for 'onlineSavingsMsg'
Learn More

DL-1-Phenylethylamine 98.0+%, TCI America™

Supplier:  TCI America M0165100ML

 View more versions of this product

Catalog No. M0165100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

DL-1-Phenylethylamine
-65°C
185°C
MFCD00008069
2922
RQEUFEKYXDPUSK-UHFFFAOYSA-N
1-phenylethanamine
7408
121.18
Liquid
618-36-0
Yellow
C8H11N
100 mL
1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
CC(C1=CC=CC=C1)N
121.183
CHEBI:670
≥98.0% (GC,T)

Chemical Identifiers

618-36-0
121.183
RQEUFEKYXDPUSK-UHFFFAOYSA-N
7408
1-phenylethanamine
C8H11N
MFCD00008069
1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
CHEBI:670
CC(C1=CC=CC=C1)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2692

RTECSNumber : DP5775000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions