missing translation for 'onlineSavingsMsg'
Learn More

DL-1-Phenylethylamine 98.0+%, TCI America™

Catalog No. M0165500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0165500ML 500 mL
M016525ML 25 mL
M0165100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0165500ML Supplier TCI America Supplier No. M0165500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 618-36-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008069
InChI Key RQEUFEKYXDPUSK-UHFFFAOYSA-N
Synonym 1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
PubChem CID 7408
ChEBI CHEBI:670
IUPAC Name 1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -65°C
Color Yellow
Boiling Point 185°C
Quantity 500 mL
UN Number 2922
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material DL-1-Phenylethylamine
EINECSNumber (3)-2692
RTECSNumber DP5775000
TSCA Yes