missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-1-Amino-2-propanol 98.0+%, TCI America™
$69.31 - $294.56
Chemical Identifiers
| CAS | 78-96-6 |
|---|---|
| Molecular Formula | C3H9NO |
| Molecular Weight (g/mol) | 75.11 |
| MDL Number | MFCD00008139 |
| InChI Key | HXKKHQJGJAFBHI-UHFFFAOYNA-N |
| Synonym | 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine |
| PubChem CID | 4 |
| ChEBI | CHEBI:19030 |
| IUPAC Name | 1-aminopropan-2-ol |
| SMILES | CC(O)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A122925ML
|
TCI America
A122925ML |
25 mL |
Each for $69.31
|
|
|||||
|
A1229500ML
|
TCI America
A1229500ML |
500 mL |
Each for $294.56
|
|
|||||
Chemical Identifiers
| 78-96-6 | |
| 75.11 | |
| HXKKHQJGJAFBHI-UHFFFAOYNA-N | |
| 4 | |
| 1-aminopropan-2-ol |
| C3H9NO | |
| MFCD00008139 | |
| 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine | |
| CHEBI:19030 | |
| CC(O)CN |
Specifications
| 78-96-6 | |
| Yellow | |
| C3H9NO | |
| 25 mL | |
| 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine | |
| CC(O)CN | |
| 75.11 | |
| CHEBI:19030 | |
| ≥98.0% (GC,T) | |
| DL-1-Amino-2-propanol |
| -2°C | |
| 160°C | |
| MFCD00008139 | |
| 2735 | |
| HXKKHQJGJAFBHI-UHFFFAOYNA-N | |
| 1-aminopropan-2-ol | |
| 4 | |
| 75.11 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0323
RTECSNumber : UA5775000
TSCA : Yes