missing translation for 'onlineSavingsMsg'
Learn More

DL-1-Amino-2-propanol 92.0+%, TCI America™

Catalog No. a0406500g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
A0406500G 500 g
A040625G 25 g
2 options

Catalog No. A0406500G

Supplier: TCI America A0406500G

Only null left

Chemical Identifiers

CAS 78-96-6
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008139
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine
PubChem CID 4
ChEBI CHEBI:19030
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 160°C
Quantity 500 g
UN Number 2735
Formula Weight 75.11
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-1-Amino-2-propanol
EINECSNumber (2)-0323
RTECSNumber UA5775000
TSCA Yes