missing translation for 'onlineSavingsMsg'
Learn More

DL-1-Amino-2-propanol 92.0+%, TCI America™

Catalog No. A040625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A040625G 25 g
A0406500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A040625G Supplier TCI America Supplier No. A040625G

Chemical Identifiers

CAS 78-96-6
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008139
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym 1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine
PubChem CID 4
ChEBI CHEBI:19030
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 2735
Formula Weight 75.11
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-1-Amino-2-propanol
EINECSNumber (2)-0323
RTECSNumber UA5775000
TSCA Yes