missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. p-124302
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327190010 1 g Glass Bottle
AC327190100 10 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327190010 Supplier Thermo Scientific Chemicals Supplier No. 327190010
Only null left

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12-3-3483
ChEBI CHEBI:42170

Specifications

Absorbance (0.1 M in water), (0.1 M in water) at 0nm, 0.1 max. at 280nm, 0.5 max. at 260nm
Melting Point 39.0°C to 44.0°C
Color Colorless to Yellow
Boiling Point 125.0°C to 130.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Molecular Formula C4H10O2S2
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Quantity 1 g
MDL Number MFCD00004877
Beilstein 01, III, 2360
Merck Index 15, 3419
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES C(C(C(CS)O)O)S
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Molecular Weight (g/mol) 154.24
PubChem CID 446094
Formula Weight 154.24
Percent Purity ≥99%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material DL-1, 4-Dithiothreitol, DNase, RNase, Protease free
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-531-9
RTECSNumber XO8576500

RUO – Research Use Only