missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 98%, Pure

Catalog No. AC409190010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409190010 1 g Glass bottle
AC409190050 5 g Glass bottle
AC409190250 25 g Glass bottle
AC409191000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC409190010 Supplier Thermo Scientific Chemicals Supplier No. 409190010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3483-12-3 | C4H10O2S2 | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.25
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Melting Point 39.0°C to 44.0°C
Color White
Boiling Point 125.0°C to 130.0°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CH(OH)CH(OH)CH2SH
Quantity 1 g
Beilstein 01, III, 2360
Merck Index 15, 3419
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in ethanol,acetone,ether,,chloroform and ethyl acetate
Formula Weight 154.25
Percent Purity 98%
Grade Pure
Physical Form Crystalline Solid
Chemical Name or Material DL-1, 4-Dithiothreitol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-468-7

RUO – Research Use Only