missing translation for 'onlineSavingsMsg'
Learn More

DL-1,4-Dichloro-2,3-butanediol 98.0+%, TCI America™

Catalog No. p-7127725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34235G 5 g
D342325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34235G Supplier TCI America Supplier No. D34235G
Only null left

Chemical Identifiers

CAS 2419-73-0
Molecular Formula C4H8Cl2O2
Molecular Weight (g/mol) 159.006
InChI Key SAUBRJOIKMVSRU-UHFFFAOYSA-N
PubChem CID 102809
IUPAC Name 1,4-dichlorobutane-2,3-diol
SMILES C(C(C(CCl)O)O)Cl

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 159.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material DL-1,4-Dichloro-2,3-butanediol