missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-1,4-Dichloro-2,3-butanediol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 77°C |
| Color | White-Yellow |
| Boiling Point | 152°C |
| Quantity | 5 g |
| Formula Weight | 159.01 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-1,4-Dichloro-2,3-butanediol |
Safety and Handling
| TSCA | Yes |