missing translation for 'onlineSavingsMsg'
Learn More

DL-1,2-Hexanediol, 98%

Catalog No. AC200132500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC200132500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC200132500 Supplier Thermo Scientific Chemicals Supplier No. 200132500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6920-22-5 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6920-22-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00010737
InChI Key FHKSXSQHXQEMOK-LURJTMIESA-N
Synonym 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol
PubChem CID 94335
IUPAC Name hexane-1,2-diol
SMILES CCCC[C@H](O)CO

Specifications

Melting Point -25.0°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 223.0°C to 224.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430
Linear Formula CH3(CH2)3CH(OH)CH2OH
Quantity 250 g
Beilstein 01, I, 251
Specific Gravity 0.95
Solubility Information Solubility in water: soluble.
Viscosity 87 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-1, 2-Hexanediol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 230-029-6

RUO – Research Use Only