missing translation for 'onlineSavingsMsg'
Learn More

DL-1-(1-Naphthyl)ethylamine 98.0+%, TCI America™

Catalog No. N052325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N052325ML 25 mL
N0523500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N052325ML Supplier TCI America Supplier No. N052325ML

Chemical Identifiers

CAS 42882-31-5
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00004014
InChI Key RTCUCQWIICFPOD-UHFFFAOYSA-N
Synonym +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine
PubChem CID 98089
IUPAC Name 1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 mL
Formula Weight 171.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material DL-1-(1-Naphthyl)ethylamine
EINECSNumber (4)-1343
TSCA Yes