missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Divinylbenzene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$380.51 - $380.51
Specifications
| CAS | 1321-74-0 |
|---|---|
| Density | 0.914 g/mL (at 25°C) |
| Boiling Point | 195°C |
| Molecular Formula | C10H10 |
| Linear Formula | C6H4(CH=CH2)2 |
Description
Divinylbenzene (DVB) is a bis unsaturated monomer. DVB along with styrene as a co monomer is used in producing cross linked polymers. Several examples have reported. Polymerization with high concentrations of DVB produces popcorn polymers.DVB is used in polymeric core shell nanoparticles and as a crosslinking agent. DVB may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.Specifications
| 1321-74-0 | |
| 195°C | |
| C6H4(CH=CH2)2 | |
| MFCD01778656 | |
| 130.19 |
| 0.914 g/mL (at 25°C) | |
| C10H10 | |
| n20/D 1.561 (literature) | |
| Diethenylbenzene | |
| 80% |