missing translation for 'onlineSavingsMsg'
Learn More

Divinyl Sulfone (stabilized with HQ) 96.0+%, TCI America™

Catalog No. D09595G
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09595G 5 g
D095925G 25 g
D0959250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D09595G Supplier TCI America Supplier No. D09595G

Chemical Identifiers

CAS 77-77-0
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00008623
InChI Key AFOSIXZFDONLBT-UHFFFAOYSA-N
Synonym divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
PubChem CID 6496
ChEBI CHEBI:53729
IUPAC Name 1-ethenylsulfonylethene
SMILES C=CS(=O)(=O)C=C

Specifications

Melting Point -26°C
Boiling Point 234°C
Quantity 5 g
UN Number 2927
Formula Weight 118.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Divinyl Sulfone (stabilized with HQ)
RTECSNumber KM7175000
TSCA No
Recommended Storage Refrigerator