missing translation for 'onlineSavingsMsg'
Learn More

Divinyl sulfone, 97%, stab. with 0.05% hydroquinone

Catalog No. AAL1282718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1282718 50 g
AAL1282709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1282718 Supplier Thermo Scientific Chemicals Supplier No. L1282718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77-77-0 | C4H6O2S | 118.15 g/mol

Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.

Chemical Identifiers

CAS 77-77-0
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00008623
InChI Key AFOSIXZFDONLBT-UHFFFAOYSA-N
Synonym divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
PubChem CID 6496
ChEBI CHEBI:53729
IUPAC Name 1-ethenylsulfonylethene
SMILES C=CS(=O)(=O)C=C

Specifications

Melting Point ∼-26°C
Density 1.177
Boiling Point 233°C to 234°C
Flash Point 102°C (215°F)
Refractive Index 1.476
Quantity 50 g
UN Number UN2927
Beilstein 1071329
Solubility Information Miscible with water.
Formula Weight 118.16
Percent Purity 97%
Chemical Name or Material Divinyl sulfone, stabilized with 0.05% hydroquinone
Show More Show Less
Hazard Category H300+H310-H314-H335
Hazard Statement GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 201-057-6
RTECSNumber KM7175000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only