missing translation for 'onlineSavingsMsg'
Learn More

Divinyl Adipate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. A118825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A118825G 25 g
A1188100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A118825G Supplier TCI America Supplier No. A118825G
Only null left

Chemical Identifiers

CAS 4074-90-2
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00059202
InChI Key JZQAAQZDDMEFGZ-UHFFFAOYSA-N
Synonym Adipic Acid Divinyl Ester
PubChem CID 19997
IUPAC Name 1,6-diethenyl hexanedioate
SMILES C=COC(=O)CCCCC(=O)OC=C

Specifications

Color White
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Divinyl Adipate (stabilized with MEHQ)
EINECSNumber (2)-0870
RTECSNumber AV1800000
TSCA No