missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Divided Hearing Protection Dispenser

Catalog No. 19129483
Change view
Click to view available options
Height (English):
11.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
19129483 11.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19129483 Supplier Brady™ Supplier No. PD265E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylic dispensers serve as a visual reminder for employees to wear their ear plugs. Conveniently mount near hazardous work areas to encourage compliance.

  • White divided acrylic ear plug dispenser with clear covering
  • Text Legend Hearing Protection in black lettering
  • Surface mounted
  • 11.5 H x 15.375 W x 9 D in.

Specifications

Depth (English) 9 in.
Depth (Metric) 22.8 cm
For Use With (Application) Surface Mounted Ear Plug Dispenser to be Positioned Near Work Areas Where Hearing Protection is Required.
Height (English) 11.5 in.
Height (Metric) 29.2 cm
Holds Ear plugs
Width (Metric) 39.1 cm
Width (English) 15.4 in.
Product Type Ear plug refill box
Disposable Reusable
Material Acrylic
Mounting Flat Surface
Show More Show Less