missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Divided Eyewear Dispenser

Catalog No. 19129472
Change view
Click to view available options
Height (English):
11.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
19129472 11.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19129472 Supplier Brady™ Supplier No. PD264E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eyewear dispensers designed to be mounted near all hazardous work areas to remind employees to wear their eyewear.

  • Package includes dispenser only
  • Eyewear sold separately

Specifications

Height (English) 11.5 in.
Width (English) 15.375 in.
Color Black on White
Device Holds Approx. 20 pairs of eye glasses
Depth (English) 9 in.
For Use With (Application) Facility Identification
Material Acrylic