Learn More
Thermo Scientific™ Divalproex sodium
Supplier: Thermo Scientific™ 463490250
| Quantity | 25 g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 76584-70-8 | |
| 310.41 | |
| sodium 2-propylpentanoic acid 2-propylpentanoate |
| C16H31NaO4 | |
| MSRILKIQRXUYCT-UHFFFAOYSA-M | |
| [Na+].CCCC(CCC)C(O)=O.CCCC(CCC)C([O-])=O |
Specifications
| Divalproex sodium | |
| C16H31NaO4 | |
| Solubility aq. soln.: >100 g/L | |
| [Na+].CCCC(CCC)C(O)=O.CCCC(CCC)C([O-])=O | |
| 310.41 |
| 76584-70-8 | |
| 25 g | |
| MSRILKIQRXUYCT-UHFFFAOYSA-M | |
| sodium 2-propylpentanoic acid 2-propylpentanoate | |
| 310.4 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May damage the unborn child.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger