Learn More
Tecan Systems 350L Tips ANSI NF Nest
Catalog No. NC0725391
Encompass
Genuine Tecan 350 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, nested 5 stack, non-filtered, conductive. Tecan STERILE purity level - Sterile, tested & certified to be free from human DNA, Dnase, Rnase, pyrogens & endotoxins. 16 tip boxes x 480 tips, 7,680 tips/sales unit. LiHa Disposable Tips have been carefully designed & validated to work specifically w/ Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality & reproducibility, ensuring reliable liquid handling performance. Mfg is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures & premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera & image recognition software, checking all functional parts to ensure consistent pipetting accuracy & precision.
Catalog No. NC0725391 Supplier Tecan Systems Supplier No. 30083401
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Genuine Tecan 350 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, nested 5 stack, non-filtered, conductive. Tecan STERILE purity level - Sterile, tested & certified to be free from human DNA, Dnase, Rnase, pyrogens & endotoxins. 16 tip boxes x 480 tips, 7,680 tips/sales unit. LiHa Disposable Tips have been carefully designed & validated to work specifically w/ Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality & reproducibility, ensuring reliable liquid handling performance. Mfg is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures & premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera & image recognition software, checking all functional parts to ensure consistent pipetting accuracy & precision.