missing translation for 'onlineSavingsMsg'
Learn More

Dithizone Silver Complex 97.0+%, TCI America™

Catalog No. D16791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D16791G Supplier TCI America Supplier No. D16791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36539-86-3
Molecular Formula C13H11AgN4S
Molecular Weight (g/mol) 363.187
MDL Number MFCD00059010
InChI Key RTNZRONZKMOMBV-UHFFFAOYSA-M
PubChem CID 132770964
IUPAC Name silver;N'-anilino-N-phenyliminocarbamimidothioate
SMILES C1=CC=C(C=C1)NN=C(N=NC2=CC=CC=C2)[S-].[Ag+]

Specifications

Quantity 1 g
Formula Weight 363.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dithizone Silver Complex