missing translation for 'onlineSavingsMsg'
Learn More

Dithizone, MilliporeSigma

Catalog No. 01213173
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01213173 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01213173 Supplier Merck Emd Millipore Supplier No. DX23703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Dithizone is a high purity product that meets reagent specifications suitable for testing USP/NF monographs.

Specifications

Boiling Point Not Measured
Quantity 25 g
Molecular Weight (g/mol) Not Measured
Percent Purity Not Measured
Grade ACS
Physical Form Crystalline