missing translation for 'onlineSavingsMsg'
Learn More

Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure

Used as protective agent for SH groups | CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

Supplier:  Thermo Fisher Scientific J1539714

 View more versions of this product

Catalog No. AAJ1539714


Only null left
Explore available promotions

Description

Description

Used as a protective agent for SH groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dithiothreitol
40°C to 43°C
>110°C
C4H10O2S2
25 g
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
C(C(C(CS)O)O)S
154.24
CHEBI:42170
≥99.5%
Crystalline Powder
12-3-3483
125°C to 130°C/12 mm
Plastic bottle
MFCD00004877
1719757
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2 S,3 S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.25
Molecular Biology, Ultrapure

Chemical Identifiers

12-3-3483
154.24
VHJLVAABSRFDPM-IMJSIDKUSA-N
446094
(2 S,3 S)-1,4-bis(sulfanyl)butane-2,3-diol
C4H10O2S2
MFCD00004877
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
CHEBI:42170
C(C(C(CS)O)O)S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 222-468-7

RTECSNumber : EK1610000

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only