missing translation for 'onlineSavingsMsg'
Learn More

Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure

Catalog No. AAJ1539722
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1539722 100 g Plastic bottle
AAJ1539703 1 g Plastic bottle
AAJ1539706 5 g Plastic bottle
AAJ1539714 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ1539722 Supplier Thermo Fisher Scientific Supplier No. J1539722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as protective agent for SH groups | CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

Used as a protective agent for SH groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2 S,3 S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Melting Point 40°C to 43°C
Boiling Point 125°C to 130°C/12 mm
Flash Point >110°C
Packaging Plastic bottle
Quantity 100 g
Beilstein 1719757
Formula Weight 154.25
Percent Purity ≥99.5%
Grade Molecular Biology, Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material Dithiothreitol
Show More Show Less
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-468-7
RTECSNumber EK1610000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only