Learn More
Abcam Inc Disuccinimidyl suberate, cross-linking reagent, 5G
Catalog No. 502706725
Encompass
MW 368.34 Da, Purity >99%. Homobifunctional primary amine reactive cross-linker. Contains an 8 atom linker (11.4 ) and can cross-link distances up to ~25 due to protein flexibility. Produces no toxic side products.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502706725 Supplier Abcam Inc Supplier No. AB1412745G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 368.34 Da, Purity >99%. Homobifunctional primary amine reactive cross-linker. Contains an 8 atom linker (11.4 ) and can cross-link distances up to ~25 due to protein flexibility. Produces no toxic side products.
The product is subject to the following: Abcam Restricted Use Statement