missing translation for 'onlineSavingsMsg'
Learn More

Disuccinimidyl suberate, 97%

Catalog No. aah5177103
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5177103 1 g
AAH51771ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5177103 Supplier Thermo Scientific Chemicals Supplier No. H5177103
Only null left

CAS: 68528-80-3 | C16H20N2O8 | 368.342 g/mol

Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl suberate is a homobifunctional amine-reactive crosslinker. It is usually coupled to molecules containing primary amines via amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates an 8-atom spacer arm.

Solubility
Insoluble in water. Soluble in chloroform: 50 mg/mL.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 68528-80-3
Molecular Formula C16H20N2O8
Molecular Weight (g/mol) 368.342
MDL Number MFCD00049059
InChI Key ZWIBGKZDAWNIFC-UHFFFAOYSA-N
Synonym disuccinimidyl suberate, bis 2,5-dioxopyrrolidin-1-yl octanedioate, suberic acid bis n-hydroxysuccinimide ester, di n-succinimidyl suberate, bicl100, 2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, nhs-sa, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester
PubChem CID 100658
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) octanedioate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 166°C to 170°C
Quantity 1 g
Solubility Information Insoluble in water. Soluble in chloroform: 50mg/mL.
Formula Weight 368.34
Percent Purity 97%
Chemical Name or Material Disuccinimidyl suberate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only