Learn More
Chemglass Inc Distilling Receiver, 15mL, 14/20, Complete, Approx 210mm Height x 90mm Width
Catalog No. CG127914
Encompass
Distilling Receiver, 15mL, 14/20, Complete, Approx 210mm Height x 90mm Width. Radial style distillation receiver allows four flasks to be progressively filled. Upper vacuum adapter is bent at a 105 angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.
Catalog No. CG127914 Supplier Chemglass Inc Supplier No. CG127914
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Distilling Receiver, 15mL, 14/20, Complete, Approx 210mm Height x 90mm Width. Radial style distillation receiver allows four flasks to be progressively filled. Upper vacuum adapter is bent at a 105 angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.