Learn More
Chemglass Life Sciences Distilling Receiver, 5mL, Moisture Test, Bidwell-Sterling, 24/40 Joint Size, Approx. 275mm Height x 90mm Width DFS Item
Supplier:  Chemglass Inc CG125703
Distilling Receiver, 5mL, Moisture Test, Bidwell-Sterling, 24/40 Joint Size, Approx. 275mm Height x 90mm Width. Bidwell-Sterling type receiver used to determine the moisture in foods and organics. Made to ASTM specification E123. Also conforms with many methods of American Oil Chemists Society. Both the 5 and 10ml capacities have subdivisions of 0.1mL.
Catalog No. CG125703
Only null left
Explore available promotions