Learn More
Chemglass Inc Distilling Receiver, 250mL, 24/40, Complete
Catalog No. CG127601
Encompass
Distilling Receiver, 250mL, 24/40, Complete. Cow type design has three receivers 45 apart. Condensers with lower drip tip inner joints will protrude into the spherical section, permitting the distillate to fall directly into the receiving flasks. Distribution adapter has a serrated connection for attachment to vacuum source. Complete item supplied with distribution adapter, three flasks of the listed size and six stainless steel springs.
Catalog No. CG127601 Supplier Chemglass Inc Supplier No. CG127601
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Distilling Receiver, 250mL, 24/40, Complete. Cow type design has three receivers 45 apart. Condensers with lower drip tip inner joints will protrude into the spherical section, permitting the distillate to fall directly into the receiving flasks. Distribution adapter has a serrated connection for attachment to vacuum source. Complete item supplied with distribution adapter, three flasks of the listed size and six stainless steel springs.