Learn More
Chemglass Inc Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 325mm Height x 95mm Width
Catalog No. CG126311
Encompass
Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 325mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. CG126311 Supplier Chemglass Inc Supplier No. CG126311
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 325mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.