Learn More
Research Products Corporation Distilled Water, 55 Gal
Catalog No. 502230234
Encompass
Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.
Catalog No. 502230234 Supplier Research Products Corporation Supplier No. W2052555
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.