Research Products Corporation Distilled Water, Laboratory Reagent Grade, 4 Liters
Catalog No. 50712782
Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.
Catalog No.
50712782
Supplier
Research Products CorporationSupplier No.
RPI W205254000.0
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.