Learn More
Chemglass Life Sciences Distilling Head, 24/40 Joint Size, 250mm Condenser Length, Approx. 165mm Height x 420mm Width
Supplier: Chemglass Inc CG123807
Distilling Head, 24/40 Joint Size, 250mm Condenser Length, Approx. 165mm Height x 420mm Width. Simple distillation head with a condenser sealed to the adapter at a 75 angle. The sidearm is sealed at a height for use with a 1 immersion thermometer. The 24/40 and 29/42 sizes are made to accept a 10/30, 3 immersion thermometer. The inner drip tip joint on the condenser and inner joint on the uptake tube are the same size.