missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Dissecting Needle Wood

Sharp-pointed, dissecting needle with wood holder.

$236.17

Specifications

Product Type Needle
Autoclavable Autoclavable
Color Wood
Material Stainless Steel
For Use With (Application) Dissection
View More Specs

Products 1
Catalog Number Tip Style Price Quantity & Availability  
Catalog Number Tip Style Price Quantity & Availability  
13820024
View Documents Promotion Details
Straight
Pack of 12 Each for $236.17
Case of 12 Pack for $2,834.04
Only null left
 
Description

Description

Available in straight or bent formats, all devices have a needle length of 45mm (1.75 in.).

  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Specifications

Needle
Wood
Dissection
5.75 in.
145 mm
N/A
N/A
Autoclavable
Stainless Steel
Dissecting Needles
5.75 in.
N/A
10 g
Videos
SDS
Documents

Documents