Learn More
Mopec™ Dissecting Board
Mopec Dissecting Board is ideal for pathology specimen dissection
Supplier:  Mopec™ BC031
 View more versions of this product
Catalog No. 22444322
Only null left
Description
Description
  • Easy to clean
  • Safe, rounded edges
  • Ergonomic, multi-user work surface
Specifications
Chemical Identifiers
Dissecting Board
Polyethylene
Specifications
Light Blue
Dissecting Board
Polyethylene
23 x 16 in.
Product Suggestions
Product Suggestions