Learn More
Mopec™ Dissecting Board with 12-inch Legs
Catalog No. p-7116289
Change view
Click to view available options
Color:
Dark Blue
Light Blue
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22444326 White
BC060 Light Blue
22444338 Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22444326 Supplier Mopec™ Supplier No. BC050
Only null left
Mopec Dissecting Board with 12-inch Legs is ideal for pathology specimen dissection
  • Easy to clean
  • Safe, rounded edges
  • Ergonomic, multi-user work surface
  • Optional removable legs
  • Non-skid feet
Specifications
Product Type Dissecting Board
Color White
Description 12-inch legs
Material Polyethylene
Dimensions (L x W) 23 x 16 in.