Learn More
Mopec™ Dissecting Board with 12-inch Legs
Catalog No. BC060
Change view
Click to view available options
Color:
Dark Blue
Light Blue
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
BC060 Light Blue
22444326 White
22444338 Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BC060 Supplier Mopec™ Supplier No. BC060
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mopec Dissecting Board with 12-inch Legs is ideal for pathology specimen dissection
  • Easy to clean
  • Safe, rounded edges
  • Ergonomic, multi-user work surface
  • Optional removable legs
  • Non-skid feet
Specifications
Product Type Dissecting Board
Color Light Blue
Description 12-inch legs
Material Polyethylene
Dimensions (L x W) 23 x 16 in.