Learn More
Mopec™ Dissecting Boards with 3-inch Legs
Mopec Dissecting Boards are ideal for a wide variety of pathology applications including photography, specimen isolation, specimen pinning and satellite operations.
Supplier:  Mopec™ BC068
 View more versions of this product
Catalog No. 22444337
Only null left
Description
Description
  • Easy to clean
  • Safe, rounded edges
  • Ergonomic, multi-user work surface
  • Optional removable legs
  • Non-skid feet
Specifications
Chemical Identifiers
Dissecting Board
Polyethylene
Specifications
Dark Blue
Dissecting Board
Polyethylene
16 x 12 in.
Product Suggestions
Product Suggestions