Learn More
FEATHER Dissecting Blade Handle F-60
Catalog No. 0892425A
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
0892425A 1 Pk.
1 options
Catalog No. 0892425A Supplier FEATHER Supplier No. F 60
FEATHER Dissecting Blades are designed for autopsy. The extra-thick stainless steel blade provides high rigidity and extra-long life. Can be used with autoclavable handle designed for easy blade replacement and cleaning.
  • Made in Japan
Specifications
Blade Material Special Resin (Autoclavable)
Product Type Dissecting Handle
Length (Metric) 145 mm
For Use With (Equipment) FEATHER Dissecting Blades No. 61 or No. 62
Quantity 1 Pk.