Learn More
FEATHER Dissecting Blade
Catalog No. 0892425C
Change view
Click to view available options
Product Type:
No. 61 (Wide blade)
No. 62 (Narrow blade)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
0892425C No. 62 (Narrow blade)
0892425B No. 61 (Wide blade)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0892425C Supplier FEATHER Supplier No. 62
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FEATHER Dissecting Blades are designed for autopsy. The extra-thick stainless steel blade provides high rigidity and extra-long life. Can be used with autoclavable handle designed for easy blade replacement and cleaning.
  • Made in Japan
Specifications
Blade Material Stainless Steel
Product Type No. 62 (Narrow blade)
Length (Metric) 77 mm
For Use With (Equipment) FEATHER Dissecting Blade Handle F-60
Quantity 20 Pk.
Width (Metric) 0.6 mm