Waters Limited DisQuE 1.5 g Sodium Acetate and 6 g MgSO4, 50 mL Tube, 100/pk
Catalog No. 50791149
This is tube contains the salts required for the extraction step in QUECHERS AOAC Method 2007.01. QUECHERS (pronounced catchers) methods are used for multi-pesticide residue analyses in food. This is a 100 pack of tubes containing the salts.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This is tube contains the salts required for the extraction step in QUECHERS AOAC Method 2007.01. QUECHERS (pronounced catchers) methods are used for multi-pesticide residue analyses in food. This is a 100 pack of tubes containing the salts.