Learn More
Waters Limited DisQuE 150 mg MgSO4, 25 mg PSA & 25 mg C18, 2 mL Tube, 100/pk
Catalog No. 50791210
Encompass
This is clean-up tube is used for the extract from step 1 in QUECHERS CEN Method 15662. This d-SPE tube contains C18 for use with high fat food samples. QUECHERS (pronounced catchers) methods are used for multi-pesticide residue analyses in food . This is a 100 pack of tubes containing the d-SPE components.
Catalog No. 50791210 Supplier Waters Limited Supplier No. 186004832
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This is clean-up tube is used for the extract from step 1 in QUECHERS CEN Method 15662. This d-SPE tube contains C18 for use with high fat food samples. QUECHERS (pronounced catchers) methods are used for multi-pesticide residue analyses in food . This is a 100 pack of tubes containing the d-SPE components.