Learn More
Fisherbrand™ Disposable SMS Laboratory Coats
Catalog No. 14901011
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
14901011 Blue 2X-Large
14901001 White Medium
14901002 White Large
14901003 White X-Large
14901004 White 2X-Large
14901005 White 3X-Large
14901006 White 5X-Large
14901007 Blue Small
14901008 Blue Medium
14901009 Blue Large
14901010 Blue X-Large
14901012 Blue 3X-Large
14901013 Blue 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 14901011 Supplier Fisherbrand™ Supplier No. 14901011
Only null left
Fisherbrand™ Disposable SMS Lab Coats come complete with a knitted collar and cuffs. Designed to protect healthcare professionals, lab technicians and other users from fluids and cross-contamination.
Fisherbrand™ premium protective apparel is available in both lab coats and hip length jackets. Made from high quality, ultra-soft SMS fabric designed to keep you protected and comfortable.
  • Fluid-resistant material meets AAMI L2 standards
  • 42 GSM high quality, breathable SMS material
  • Anti-static coating
  • High quality front snap button closure
  • Comfortable knit collar and cuffs
  • 3-pocket design
  • Convenient side slits for easy access to pockets
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
Style Knee-length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Certifications/Compliance ANSI, AAMI PB70
Packaging Type Bagged
Autoclavable Not Autoclavable
No. per Pack 10/Pk.
Show More Show Less